missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dinitrobenzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1522306

 View more versions of this product

Catalog No. AAA1522306



Specifications

Specifications

3,5-Dinitrobenzonitrile
5g
C7H3N3O4
UN3439
SSDNULNTQAUNFQ-UHFFFAOYSA-N
3,5-dinitrobenzonitrile
20062
98%
127°C to 130°C
4110-35-4
MFCD00007228
benzonitrile, 3,5-dinitro, 3,5-dinitro-benzonitrile, 3,5-dinitrobenzenecarbonitrile, acmc-209jih, 3,5-bisnitrobenzonitrile, 3,5-dinitrobenzonitrile, wln: wnr cnw ecn, benzonitrile,3,5-dinitro, 1,3-dinitro-5-cyanobenzene, 5-cyano-1,3-dinitrobenzene
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C#N
193.118
193.12

Chemical Identifiers

4110-35-4
193.118
SSDNULNTQAUNFQ-UHFFFAOYSA-N
20062
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C#N
C7H3N3O4
MFCD00007228
benzonitrile, 3,5-dinitro, 3,5-dinitro-benzonitrile, 3,5-dinitrobenzenecarbonitrile, acmc-209jih, 3,5-bisnitrobenzonitrile, 3,5-dinitrobenzonitrile, wln: wnr cnw ecn, benzonitrile,3,5-dinitro, 1,3-dinitro-5-cyanobenzene, 5-cyano-1,3-dinitrobenzene
3,5-dinitrobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : DI4365000

Documents