missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dinitrobenzoic acid, 98+%, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC153191000

 View more versions of this product

Catalog No. AC153191000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 5-Dinitrobenzoic acid
Plastic bottle
205.0°C to 209.0°C
99%
98.5
98% min. (HPLC)
(O2N)2C6H3CO2H
09,413
3,5-dinitro-benzoic acid
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
212.12
CHEBI:73914
98+%
1.6830g/mL
1.683
100g
99-34-3
100.0
C7H4N2O6
MFCD00007253
15,3302
Solubility in water: 1350mg/l (25°C). Other solubilities: very soluble in alcohol and glacial acetic acid,sparingly soluble in ether,benzene and,carbon disulfide
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)O
7433
212.12

Chemical Identifiers

99-34-3
212.12
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
7433
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)O
C7H4N2O6
MFCD00007253
3,5-dinitro-benzoic acid
CHEBI:73914
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 202-751-1

RTECSNumber : DG9140700

TSCA : TSCA

SDS Documents