Learn More
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
$87.75 - $305.19
Chemical Identifiers
CAS | 99-34-3 |
---|---|
Molecular Formula | C7H4N2O6 |
Molecular Weight (g/mol) | 212.12 |
MDL Number | MFCD00007253 |
InChI Key | VYWYYJYRVSBHJQ-UHFFFAOYSA-N |
Synonym | 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid |
PubChem CID | 7433 |
ChEBI | CHEBI:73914 |
IUPAC Name | 3,5-dinitrobenzoic acid |
SMILES | OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1650230
|
Thermo Scientific Chemicals
A1650230 |
250 g |
Each for $87.75
|
|
|||||
AAA165020B
|
Thermo Scientific Chemicals
A165020B |
1000 g |
Each for $305.19
|
|
|||||
Description
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
Solubility
It is Soluble in ethanol (50 mg/mL), methanol, alcohol (very), and glacial acetic acid (very). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases, Strong reducing agents.
Chemical Identifiers
99-34-3 | |
212.12 | |
VYWYYJYRVSBHJQ-UHFFFAOYSA-N | |
7433 | |
3,5-dinitrobenzoic acid |
C7H4N2O6 | |
MFCD00007253 | |
3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid | |
CHEBI:73914 | |
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
Specifications
99-34-3 | |
1.68 | |
MFCD00007253 | |
UN1325 | |
14,3276 | |
It is Soluble in ethanol (50mg/mL),methanol,alcohol (very),and glacial acetic acid (very). Insoluble in water. | |
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O | |
212.12 | |
CHEBI:73914 | |
≥98% |
204°C to 208°C | |
C7H4N2O6 | |
250 g | |
1914286 | |
3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid | |
VYWYYJYRVSBHJQ-UHFFFAOYSA-N | |
3,5-dinitrobenzoic acid | |
7433 | |
212.12 | |
3,5-Dinitrobenzoic acid |
Safety and Handling
GHS H Statement
H301-H228-H315-H319-H335
Toxic if swallowed.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-751-1
RTECSNumber : DG9140700
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only