missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dinitrobenzamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2186522

 View more versions of this product

Catalog No. AAB2186522



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dinitrobenzamide
181°C to 183°C
MFCD00007985
14,6612
UUKWKUSGGZNXGA-UHFFFAOYSA-N
3,5-dinitrobenzamide
4511
98%
121-81-3
C7H5N3O5
100 g
nitromide, tristat, unistat, benzamide, 3,5-dinitro, nitromide usan, component of tristat, nitroamide, component of unistat-3, unii-9duj3cmk8s, 9duj3cmk8s
NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
211.13
211.13

Chemical Identifiers

121-81-3
211.13
UUKWKUSGGZNXGA-UHFFFAOYSA-N
4511
NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
C7H5N3O5
MFCD00007985
nitromide, tristat, unistat, benzamide, 3,5-dinitro, nitromide usan, component of tristat, nitroamide, component of unistat-3, unii-9duj3cmk8s, 9duj3cmk8s
3,5-dinitrobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 204-499-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only