Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dinitro-L-tyrosine, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Dinitro-L-tyrosine
CAS 17360-11-1
Formula Weight 271.18
MDL Number MFCD00038093
Synonym 3,5-Dinitro-Tyr-OH; (S)-2-Amino-3-(4-hydroxy-3,5-dinitrophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6326503

  SDS
alfa aesar™
H6326503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6326506

  SDS
alfa aesar™
H6326506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63265MD

  SDS
alfa aesar™
H63265MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.18
Color Red
MDL Number MFCD00038093
Physical Form Powder
Assay 98%
CAS 17360-11-1
Quantity 1g
Chemical Name or Material 3,5-Dinitro-L-tyrosine
Synonym 3,5-Dinitro-Tyr-OH; (S)-2-Amino-3-(4-hydroxy-3,5-dinitrophenyl)propionic acid