missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dinitro-L-tyrosine, 98%

Manufacturer:  Alfa Aesar™ H63265MD

 View more versions of this product

Catalog No. AAH63265MD



Specifications

Specifications

3,5-Dinitro-L-tyrosine
C9H9N3O7
3,5-dinitro-l-tyrosine, 3,5-dinitrotyrosine, 3,5-dinitro-tyr-oh, s-2-amino-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, h-tyr 3,5-no2 2-oh, tyrosine, 3,5-dinitro, 2s-2-amino-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, l-tyrosine,3,5-dinitro, sazosdsflrxrea-yfkpbyrvsa, 3,5-dinitro-4-hydroxy-l-phenylalanine
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(C(=O)O)N
439948
271.18
98%
250mg
17360-11-1
MFCD00038093
SAZOSDSFLRXREA-YFKPBYRVSA-N
271.185
CHEBI:28275
Powder
Red

Chemical Identifiers

17360-11-1
271.185
SAZOSDSFLRXREA-YFKPBYRVSA-N
439948
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(C(=O)O)N
C9H9N3O7
MFCD00038093
3,5-dinitro-l-tyrosine, 3,5-dinitrotyrosine, 3,5-dinitro-tyr-oh, s-2-amino-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, h-tyr 3,5-no2 2-oh, tyrosine, 3,5-dinitro, 2s-2-amino-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, l-tyrosine,3,5-dinitro, sazosdsflrxrea-yfkpbyrvsa, 3,5-dinitro-4-hydroxy-l-phenylalanine
CHEBI:28275
SDS Documents