missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dinitro-L-tyrosine, 98%

Manufacturer:  Alfa Aesar™ H6326506

 View more versions of this product

Catalog No. AAH6326506


 

Specifications

Specifications

3,5-Dinitro-L-tyrosine
98%
MFCD00038093
SAZOSDSFLRXREA-YFKPBYRVSA-N
271.185
CHEBI:28275
Powder
5g
17360-11-1
C9H9N3O7
3,5-Dinitro-Tyr-OH; (S)-2-Amino-3-(4-hydroxy-3,5-dinitrophenyl)propionic acid
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(C(=O)O)N
439948
271.18
Red

Chemical Identifiers

17360-11-1
271.185
SAZOSDSFLRXREA-YFKPBYRVSA-N
439948
C9H9N3O7
MFCD00038093
3,5-Dinitro-Tyr-OH; (S)-2-Amino-3-(4-hydroxy-3,5-dinitrophenyl)propionic acid
CHEBI:28275
SDS Documents