Please login to your online account to display your discounted pricing

3,5-Dinitro-4-toluic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC373180050

 View more versions of this product

Catalog No. AC373180050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 226.14
Quantity 5g
Color Yellow
Melting Point 155° to 159°C
CAS Max % 100.0
Linear Formula (ON)C6H(CH3)COH
Synonym 3, 5-Dinitro-4-methylbenzoic acid
Infrared Spectrum Authentic
Chemical Name or Material 3, 5-Dinitro-4-toluic acid
Assay Percent Range 98%
Molecular Formula C8H6N2O6
Packaging Glass bottle
MDL Number MFCD00007175
CAS 16533-71-4
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable solid.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Dinitro-4-toluic acid
CAS 16533-71-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H6N2O6
Linear Formula (ON)C6H(CH3)COH
Formula Weight 226.14
MDL Number MFCD00007175
Synonym 3, 5-Dinitro-4-methylbenzoic acid
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable solid.

SDS & Safety Documents