missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethylthiophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 259230025

 View more versions of this product

Catalog No. AC259230025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 5-Dimethylthiophenol
Authentic
1.5665 to 1.5685 (20°C, 589nm)
2.5mL
38360-81-5
100.0
C8H10S
MFCD00040226
CESBAYSBPMVAEI-UHFFFAOYSA-N
3,5-dimethylbenzenethiol
117548
Liquid
1.02g/mL
Glass bottle
Yellow
98%
97.5
98%
CH3(C6H3)CH3SH
3,5-dimethyl thiophenol, 3,5-dimethylbenzene-1-thiol, 3,5-dimethylthiophenol, 3,5-xylenethiol, 3,5dimethylthiophenol, 5-mercapto-m-xylene, benzenethiol, 3,5-dimethyl, m-xylene-5-thiol, pubchem6885, xylenethiol, mixed isomers
CC1=CC(=CC(=C1)S)C
138.228
138.23
≥97.5%

Chemical Identifiers

38360-81-5
138.228
CESBAYSBPMVAEI-UHFFFAOYSA-N
117548
CC1=CC(=CC(=C1)S)C
C8H10S
MFCD00040226
3,5-dimethyl thiophenol, 3,5-dimethylbenzene-1-thiol, 3,5-dimethylthiophenol, 3,5-xylenethiol, 3,5dimethylthiophenol, 5-mercapto-m-xylene, benzenethiol, 3,5-dimethyl, m-xylene-5-thiol, pubchem6885, xylenethiol, mixed isomers
3,5-dimethylbenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 253-901-8

TSCA : TSCA

SDS Documents