Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethylpyridine, 98+%

Chemical Identifiers

Chemical Name or Material 3,5-Dimethylpyridine
CAS 591-22-0
Formula Weight 107.16
MDL Number MFCD00006404
UN Number UN1993
Synonym 3,5-Lutidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36714AD

 
alfa aesar™
36714AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36714AK

 
alfa aesar™
36714AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 107.16
Refractive Index 1.5040
Physical Form Liquid
CAS 591-22-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 169° to 170°C
Chemical Name or Material 3,5-Dimethylpyridine
TSCA TSCA
Assay ≥98%
MDL Number MFCD00006404
UN Number UN1993
Synonym 3,5-Lutidine
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density 0.939g/mL
Melting Point −9°C
Quantity 50mL
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.