missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethylpyrazole, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC116705000

 View more versions of this product

Catalog No. AC116705000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3,5-Dimethylpyrazole
Plastic bottle
White
500g
98.5
99%
23,74
Solubility in water: soluble
CC1=CC(=NN1)C
96.13
96.13
99%
Authentic
218.0°C
106.0°C to 109.0°C
67-51-6
100.0
C5H8N2
1h-pyrazole, 3,5-dimethyl, 3,5 dmp, 3,5-dimethyl pyrazole, 3,5-dimethylpyrazole, 3,5-dwumetylopirazolu, 3,5-dwumetylopirazolu polish, pubchem13256, pyrazole, 3,5-dimethyl, pyrazole,5-dimethyl, unii-h21n865k9j
SDXAWLJRERMRKF-UHFFFAOYSA-N
3,5-dimethyl-1H-pyrazole
6210
Crystalline Powder or Crystals

Chemical Identifiers

67-51-6
96.13
1h-pyrazole, 3,5-dimethyl, 3,5 dmp, 3,5-dimethyl pyrazole, 3,5-dimethylpyrazole, 3,5-dwumetylopirazolu, 3,5-dwumetylopirazolu polish, pubchem13256, pyrazole, 3,5-dimethyl, pyrazole,5-dimethyl, unii-h21n865k9j
3,5-dimethyl-1H-pyrazole
C5H8N2
SDXAWLJRERMRKF-UHFFFAOYSA-N
6210
CC1=CC(=NN1)C
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to

GHS Signal Word: Warning

EINECSNumber : 200-657-5

Documents