Please login to your online account to display your discounted pricing

3,5-Dimethylpyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC399330050

 View more versions of this product

Catalog No. AC399330050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 222.09
Quantity 5g
CAS 857530-80-4
Chemical Name or Material 3, 5-Dimethylpyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C11H19BN2O2
MDL Number MFCD05663852

Specifications

Chemical Name or Material 3, 5-Dimethylpyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C11H19BN2O2
Formula Weight 222.09
MDL Number MFCD05663852