Please login to your online account to display your discounted pricing

3,5-Dimethylpyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethylpyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C11H19BN2O2
Formula Weight 222.09
MDL Number MFCD05663852
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399330010

  SDS
thermo fisher scientific
AC399330010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399330050

 
thermo fisher scientific
AC399330050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 222.09
Quantity 1g
Chemical Name or Material 3, 5-Dimethylpyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C11H19BN2O2
MDL Number MFCD05663852
CAS 857530-80-4