missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethylphenyl isothiocyanate, 99%

Manufacturer:  Alfa Aesar™ A1130214

 View more versions of this product

Catalog No. AAA1130214



Specifications

Specifications

3,5-Dimethylphenyl isothiocyanate
99%
MFCD00041082
6856812
DSMXCADWIFIJEX-UHFFFAOYSA-N
1-isothiocyanato-3,5-dimethylbenzene
142406
40046-30-8
C9H9NS
UN2922
25g
CC1=CC(=CC(=C1)N=C=S)C
163.238
163.24

Chemical Identifiers

40046-30-8
163.238
DSMXCADWIFIJEX-UHFFFAOYSA-N
1-isothiocyanato-3,5-dimethylbenzene
C9H9NS
MFCD00041082
142406
CC1=CC(=CC(=C1)N=C=S)C
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

SDS Documents