Please login to your online account to display your discounted pricing

3,5-Dimethylisoxazole 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  116381000

 View more versions of this product

Catalog No. AC116381000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 97.12
Physical Form Liquid
Quantity 100g
Color Clear colorless to pale yellow
RTECS Number NY2774200
EINECS Number 206-100-2
TSCA TSCA
Density 0.9900g/mL
Beilstein 27, 17
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003156
Chemical Name or Material 3, 5-Dimethylisoxazole
Name Note 99%
CAS 300-87-8
Assay Percent Range 99%
Molecular Formula C5H7NO

Specifications

Chemical Name or Material 3, 5-Dimethylisoxazole
Name Note 99%
CAS 300-87-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H7NO
Formula Weight 97.12
MDL Number MFCD00003156
Accessories

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents