Please login to your online account to display your discounted pricing

3,5-Dimethylisoxazole 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  116380250

 View more versions of this product

Catalog No. AC116380250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 97.12
Physical Form Liquid
Assay ≥98.5%
Quantity 25g
Color Clear colorless to pale yellow
Beilstein 27, 17
RTECS Number NY2774200
TSCA TSCA
EINECS Number 206-100-2
Density 0.9900g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
CAS Min % 98.5
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Packaging Glass bottle
Chemical Name or Material 3, 5-Dimethylisoxazole
Name Note 99%
Assay Percent Range 99%
MDL Number MFCD00003156
Molecular Formula C5H7NO
CAS 300-87-8

Specifications

Chemical Name or Material 3, 5-Dimethylisoxazole
Name Note 99%
CAS 300-87-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H7NO
Formula Weight 97.12
MDL Number MFCD00003156
Accessories

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents