Please login to your online account to display your discounted pricing

3,5-Dimethylisoxazole 99%, ACROS Organics™

$65.83 - $65.83

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethylisoxazole
Name Note 99%
CAS 300-87-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H7NO
Formula Weight 97.12
MDL Number MFCD00003156
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC116381000

  SDS
thermo fisher scientific
116381000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC116380250

  SDS
thermo fisher scientific
116380250
Glass bottle 25g Each for $65.83
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 97.12
Physical Form Liquid
Quantity 100g
Color Clear colorless to pale yellow
RTECS Number NY2774200
EINECS Number 206-100-2
TSCA TSCA
Density 0.9900g/mL
Beilstein 27, 17
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003156
Chemical Name or Material 3, 5-Dimethylisoxazole
Name Note 99%
CAS 300-87-8
Assay Percent Range 99%
Molecular Formula C5H7NO
Safety and Handling

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.