missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethylisoxazole, 99%

Manufacturer:  Alfa Aesar™ A1221714

 View more versions of this product

Catalog No. AAA1221714



Description

Description

Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

3,5-Dimethylisoxazole
142°C to 144°C
25g
99%
MFCD00003156
106324
CC1=CC(=NO1)C
97.117
97.12
31°C (87°F)
-14°C
300-87-8
C5H7NO
UN1993
FICAQKBMCKEFDI-UHFFFAOYSA-N
3,5-dimethyl-1,2-oxazole
9312

Chemical Identifiers

300-87-8
97.117
FICAQKBMCKEFDI-UHFFFAOYSA-N
3,5-dimethyl-1,2-oxazole
C5H7NO
MFCD00003156
9312
CC1=CC(=NO1)C
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : NY2774200

TSCA : TSCA

SDS Documents