missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dimethylisoxazole-4-carboxylic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1541914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,5-Dimethylisoxazole-4-carboxylic acid | |
140°C to 142°C | |
MFCD00051657 | |
115819 | |
IJEUISLJVBUNRE-UHFFFAOYSA-N | |
3,5-dimethyl-1,2-oxazole-4-carboxylic acid | |
75636 | |
99% |
2510-36-3 | |
C6H7NO3 | |
25 g | |
3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555 | |
CC1=C(C(=NO1)C)C(=O)O | |
141.126 | |
141.13 |
Chemical Identifiers
2510-36-3 | |
141.126 | |
IJEUISLJVBUNRE-UHFFFAOYSA-N | |
75636 | |
CC1=C(C(=NO1)C)C(=O)O |
C6H7NO3 | |
MFCD00051657 | |
3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555 | |
3,5-dimethyl-1,2-oxazole-4-carboxylic acid |
Safety and Handling
EINECSNumber : 219-724-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only