Please login to your online account to display your discounted pricing

3,5-Dimethylisoxazole-4-carboxylic acid, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC00601DE

 View more versions of this product

Catalog No. CC00601DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid
Quantity 5g
CAS 2510-36-3
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 141.13

Specifications

Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid
Assay Percent Range 97%
Formula Weight 141.13