Please login to your online account to display your discounted pricing

3,5-Dimethylisoxazole-4-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid
Assay Percent Range 97%
Formula Weight 141.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC00601DE

 
fisher scientific company
CC00601DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC00601EB

 
fisher scientific company
CC00601EB
Amber Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC00601DA

  SDS
fisher scientific company
CC00601DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.13
Quantity 5g
CAS 2510-36-3
Packaging Amber Glass Bottle
Assay Percent Range 97%
Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid