Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethylisoxazole-4-carbonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Dimethylisoxazole-4-carbonyl chloride
CAS 31301-45-8
Molecular Formula C6H6ClNO2
Formula Weight 159.57
MDL Number MFCD00052555
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5069503

 
alfa aesar™
H5069503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5069506

 
alfa aesar™
H5069506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.57
Chemical Name or Material 3,5-Dimethylisoxazole-4-carbonyl chloride
Assay 98%
Molecular Formula C6H6ClNO2
MDL Number MFCD00052555
CAS 31301-45-8
UN Number UN3265
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Applications
3,5-Dimethylisoxazole-4-carbonyl chloride is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with bases, oxidizing agents and moisture.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.