Please login to your online account to display your discounted pricing

3,5-Dimethylbenzenesulfonyl chloride 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391590050

 View more versions of this product

Catalog No. AC391590050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 200.64
Quantity 5g
Color White to yellow
Melting Point 91° to 95°C
CAS Min % 96.0
CAS Max % 100.0
CAS 2905-27-3
Molecular Formula C8H9ClO2S
Chemical Name or Material 3, 5-Dimethylbenzenesulfonyl chloride
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD03094655
Packaging Glass bottle
Linear Formula (CH3)2C6H3SO2Cl
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material 3, 5-Dimethylbenzenesulfonyl chloride
CAS 2905-27-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9ClO2S
Linear Formula (CH3)2C6H3SO2Cl
Formula Weight 200.64
MDL Number MFCD03094655
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents