missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethylbenzaldehyde, 98+%

Manufacturer:  Alfa Aesar™ L0974814

 View more versions of this product

Catalog No. AAL0974814



Description

Description

Applications
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

Solubility
Not miscible with water.

Notes
Handle and store under inert gas. Air Sensitive. Store away from air and oxidizing agents. Incompatible with oxidizing agents and air.
Specifications

Specifications

3,5-Dimethylbenzaldehyde
8°C to 10°C
5779-95-3
MFCD00082777
benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde
CC1=CC(=CC(=C1)C=O)C
134.178
134.18
220°C to 222°C
25g
C9H10O
2038840
NBEFMISJJNGCIZ-UHFFFAOYSA-N
3,5-dimethylbenzaldehyde
34225
≥98%

Chemical Identifiers

5779-95-3
134.178
NBEFMISJJNGCIZ-UHFFFAOYSA-N
34225
CC1=CC(=CC(=C1)C=O)C
C9H10O
MFCD00082777
benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde
3,5-dimethylbenzaldehyde
Documents