missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dimethylanisole, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115960250
Specifications
3, 5-Dimethylanisole | |
65°C | |
Glass bottle | |
0.96 | |
Colorless to Yellow | |
874-63-5 | |
100.0 | |
C9H12O | |
MFCD00008398 | |
1-methoxy-3,5-dimethyl-benzene, 3,5-dimethylanisol, 3,5-dimethylanisole, 3,5-dimethylanizole, 5-methoxy-1,3-dimethylbenzene, 5-methoxy-m-xylene, acmc-1bkxt, anisole, 3,5-dimethyl, benzene, 1-methoxy-3,5-dimethyl, ksc494e7h | |
CC1=CC(=CC(=C1)OC)C | |
136.19 | |
136.19 | |
99% |
0.9600g/mL | |
Authentic | |
1.5117 to 1.5137 | |
193°C | |
25g | |
98.5 | |
98.5% min. (GC) | |
(CH3)2C6H3OCH3 | |
06,493 | |
JCHJBEZBHANKGA-UHFFFAOYSA-N | |
1-methoxy-3,5-dimethylbenzene | |
70126 | |
Liquid |
Chemical Identifiers
874-63-5 | |
136.19 | |
JCHJBEZBHANKGA-UHFFFAOYSA-N | |
70126 | |
CC1=CC(=CC(=C1)OC)C |
C9H12O | |
MFCD00008398 | |
1-methoxy-3,5-dimethyl-benzene, 3,5-dimethylanisol, 3,5-dimethylanisole, 3,5-dimethylanizole, 5-methoxy-1,3-dimethylbenzene, 5-methoxy-m-xylene, acmc-1bkxt, anisole, 3,5-dimethyl, benzene, 1-methoxy-3,5-dimethyl, ksc494e7h | |
1-methoxy-3,5-dimethylbenzene |
Safety and Handling
EINECSNumber : 212-865-3