Please login to your online account to display your discounted pricing

3,5-Dimethylanisole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethylanisole
CAS 874-63-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
Formula Weight 136.19
MDL Number MFCD00008398
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115960250

  SDS
fisher scientific acros
115960250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC115961000

  SDS
thermo fisher scientific
AC115961000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 136.19
Quantity 25g
Color Clear colorless to light yellow
Density 96%g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.5110 to 1.5130 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
MDL Number MFCD00008398
CAS 874-63-5
Chemical Name or Material 3, 5-Dimethylanisole
Packaging Glass bottle
CAS Min % 98.5