missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyladamantane-1-methanol, 97%

Manufacturer:  Alfa Aesar™ H3784503

 View more versions of this product

Catalog No. AAH3784503


 

Specifications

Specifications

3,5-Dimethyladamantane-1-methanol
55°C to 56°C
1g
97%
MFCD00188062
RVWLWJAOIBEWAV-UHFFFAOYSA-N
(3,5-dimethyl-1-adamantyl)methanol
4050097
Solid
26919-42-6
114°C (2.25mmHg)
White
C13H22O
3,5-Dimethyl(1-hydroxymethyl)adamantane
CC12CC3CC(C1)(CC(C3)(C2)CO)C
194.318
194.32

Chemical Identifiers

26919-42-6
194.318
RVWLWJAOIBEWAV-UHFFFAOYSA-N
4050097
C13H22O
MFCD00188062
3,5-Dimethyl(1-hydroxymethyl)adamantane
SDS Documents