Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyladamantane-1-methanol, 97%

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyladamantane-1-methanol
CAS 26919-42-6
Formula Weight 194.32
MDL Number MFCD00188062
Synonym 3,5-Dimethyl(1-hydroxymethyl)adamantane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3784503

 
alfa aesar™
H3784503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3784506

 
alfa aesar™
H3784506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 55° to 56°C
Formula Weight 194.32
Chemical Name or Material 3,5-Dimethyladamantane-1-methanol
Boiling Point 114°C (2.25mmHg)
Quantity 1g
Color White
Physical Form Solid
Assay 97%
MDL Number MFCD00188062
CAS 26919-42-6
Synonym 3,5-Dimethyl(1-hydroxymethyl)adamantane