Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyladamantane-1-carboxylic acid, 97%

$93.76 - $93.76

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyladamantane-1-carboxylic acid
CAS 14670-94-1
Molecular Formula C13H20O2
Formula Weight 208.3
MDL Number MFCD00188067
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2707303

  SDS
alfa aesar™
H2707303
1g Each for $93.76
Description & Specifications

Specifications

Formula Weight 208.3
Chemical Name or Material 3,5-Dimethyladamantane-1-carboxylic acid
Assay 97%
Molecular Formula C13H20O2
Melting Point 107° to 109°C
MDL Number MFCD00188067
CAS 14670-94-1
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3,5-Dimethyladamantane-1-carboxylic acid, is used as a pharmaceutical and API intermediate.

Solubility
Sparingly Soluble (0.14 g/L) (25°C)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.