Please login to your online account to display your discounted pricing

3,5-Dimethyl-p-anisic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  187130050

 View more versions of this product

Catalog No. AC187130050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 180.2
Physical Form Crystalline Solid
Quantity 5g
Color White to brown
Melting Point 190° to 194°C
TSCA TSCA
EINECS Number 244-441-9
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Synonym 3, 5-Dimethyl-4-methoxybenzoic acid
Infrared Spectrum Authentic
Name Note 98%
Linear Formula CH3OC6H2(CH3)2CO2H
CAS 21553-46-8
Assay Percent Range 98%
MDL Number MFCD00020309
Chemical Name or Material 3, 5-Dimethyl-p-anisic acid
Molecular Formula C10H12O3

Specifications

Chemical Name or Material 3, 5-Dimethyl-p-anisic acid
Name Note 98%
CAS 21553-46-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Linear Formula CH3OC6H2(CH3)2CO2H
Formula Weight 180.2
MDL Number MFCD00020309
Synonym 3, 5-Dimethyl-4-methoxybenzoic acid
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents