missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-4H-1,2,4-triazol-4-amine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CD08995EA

 View more versions of this product

Catalog No. CD08995EA


 

Specifications Description & Specifications

Specifications

3,5-Dimethyl-4H-1,2,4-triazol-4-amine
96.5
Amber Glass Bottle
97%
C4H8N4
MFCD00051647
3530-15-2
100.0
10g
97%
112.14

Chemical Identifiers

3530-15-2
100.0
C4H8N4
MFCD00051647
97%
96.5
97%
112.14
3,5-Dimethyl-4H-1,2,4-triazol-4-amine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.