Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-4-octanone, erythro + threo, 99%

$23.66 - $23.66

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyl-4-octanone
CAS 7335-17-3
Name Note erythro + threo
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00027147
Synonym 2-Pentyl 2-butyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2079903

  SDS
alfa aesar™
B2079903
1g Each for $23.66
Description & Specifications

Specifications

Formula Weight 156.27
Name Note erythro + threo
CAS 7335-17-3
Chemical Name or Material 3,5-Dimethyl-4-octanone
Assay 99%
Molecular Formula C10H20O
MDL Number MFCD00027147
Quantity 1g
Synonym 2-Pentyl 2-butyl ketone