Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-4-nitropyridine-2-methanol, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyl-4-nitropyridine-2-methanol
CAS 149082-03-1
Molecular Formula C8H10N2O3
Formula Weight 182.18
MDL Number MFCD03426035
UN Number UN2811
Synonym 3,5-Dimethyl-2-hydroxymethyl-4-nitropyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1945206

  SDS
alfa aesar™
L1945206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1945214

  SDS
alfa aesar™
L1945214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.18
CAS 149082-03-1
MDL Number MFCD03426035
Health Hazard 1 Danger
Molecular Formula C8H10N2O3
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Melting Point 65° to 68°C
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Synonym 3,5-Dimethyl-2-hydroxymethyl-4-nitropyridine
Quantity 5g
Chemical Name or Material 3,5-Dimethyl-4-nitropyridine-2-methanol
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.