missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-4-methoxyphenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 301699-39-8
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
InChI Key WZUCSPWZHRVOSD-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-methoxyphenylboronic acid, 3,5-dimethyl-4-methoxybenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boronic acid, 4-methoxy-3,5-dimethyl-phenyl boronic acid, 4-methoxy-3,5-dimethylbenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boranediol, boronic acid, 4-methoxy-3,5-dimethylphenyl, pubchem6937, acmc-209hdl, 3,5-dimethyl-4-methoxybenzeneboronicacid
PubChem CID 3817586
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444030010 SDS View Documents Acros Organics
AC444030010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444030100 SDS View Documents Acros Organics
AC444030100
10g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

301699-39-8
180.01
3,5-dimethyl-4-methoxyphenylboronic acid, 3,5-dimethyl-4-methoxybenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boronic acid, 4-methoxy-3,5-dimethyl-phenyl boronic acid, 4-methoxy-3,5-dimethylbenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boranediol, boronic acid, 4-methoxy-3,5-dimethylphenyl, pubchem6937, acmc-209hdl, 3,5-dimethyl-4-methoxybenzeneboronicacid
C9H13BO3
WZUCSPWZHRVOSD-UHFFFAOYSA-N
3817586

Specifications

1g
95%
3,5-dimethyl-4-methoxyphenylboronic acid, 3,5-dimethyl-4-methoxybenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boronic acid, 4-methoxy-3,5-dimethyl-phenyl boronic acid, 4-methoxy-3,5-dimethylbenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boranediol, boronic acid, 4-methoxy-3,5-dimethylphenyl, pubchem6937, acmc-209hdl, 3,5-dimethyl-4-methoxybenzeneboronicacid
180.01
180.01
301699-39-8
C9H13BO3
WZUCSPWZHRVOSD-UHFFFAOYSA-N
3817586
3, 5-Dimethyl-4-methoxyphenylboronic acid