Please login to your online account to display your discounted pricing

(3,5-Dimethyl-4-isoxazolyl)methanol, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC00609DE

 View more versions of this product

Catalog No. CC00609DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 127.14
Quantity 5g
Assay 97%
CAS Max % 100.0
Molecular Formula C6H9NO2
MDL Number MFCD00464226
CAS Min % 96.5
Assay Percent Range 97%
Chemical Name or Material (3, 5-Dimethyl-4-isoxazolyl)methanol
CAS 19788-36-4

Specifications

Chemical Name or Material (3, 5-Dimethyl-4-isoxazolyl)methanol
CAS 19788-36-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9NO2
Formula Weight 127.14
MDL Number MFCD00464226