Please login to your online account to display your discounted pricing

3,5-Dimethyl-4-hydroxybenzonitrile, 97%, ACROS Organics™

$185.92 - $185.92

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzonitrile
CAS 4198-90-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00221716
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC353660050

  SDS
thermo fisher scientific
353660050
Glass bottle 5g Each for $185.92
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 147.18
Quantity 5g
Color White to beige
TSCA TSCA
RTECS Number DI4359000
Melting Point 123° to 126°C
Beilstein 10, IV, 681
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzonitrile
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H9NO
MDL Number MFCD00221716
CAS 4198-90-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic