Please login to your online account to display your discounted pricing

3,5-Dimethyl-4-hydroxybenzaldehyde, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC381850050

 View more versions of this product

Catalog No. AC381850050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 150.18
Quantity 5g
Color Tan to light brown
Melting Point 112° to 116°C
CAS Max % 100.0
Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzaldehyde
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C9H10O2
MDL Number MFCD00006946
CAS 2233-18-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Linear Formula (CH3)2C6H2(OH)CHO
Synonym 4-Hydroxy-3, 5-dimethylbenzaldehyde
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzaldehyde
CAS 2233-18-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula (CH3)2C6H2(OH)CHO
Formula Weight 150.18
MDL Number MFCD00006946
Synonym 4-Hydroxy-3, 5-dimethylbenzaldehyde
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents