Please login to your online account to display your discounted pricing

3,5-Dimethyl-4-hydroxybenzaldehyde, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzaldehyde
CAS 2233-18-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula (CH3)2C6H2(OH)CHO
Formula Weight 150.18
MDL Number MFCD00006946
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381850050

  SDS
fisher scientific acros
AC381850050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 150.18
Quantity 5g
Color Tan to light brown
Melting Point 112° to 116°C
CAS Max % 100.0
Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzaldehyde
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C9H10O2
MDL Number MFCD00006946
CAS 2233-18-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Linear Formula (CH3)2C6H2(OH)CHO
Synonym 4-Hydroxy-3, 5-dimethylbenzaldehyde
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.