Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-3-heptanol, erythro + threo, 98+%

$465.59 - $1,925.64

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyl-3-heptanol
CAS 19549-74-7
Name Note erythro + threo
Molecular Formula C9H20O
Formula Weight 144.25
MDL Number MFCD00046585
Synonym Ethyl methyl 2-methylbutyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2089618

  SDS
alfa aesar™
B2089618
50g Each for $465.59

AAB2089630

  SDS
alfa aesar™
B2089630
250g Each for $1,925.64

AAB2089609

  SDS
alfa aesar™
B2089609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3,5-Dimethyl-3-heptanol
Name Note erythro + threo
CAS 19549-74-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C9H20O
Formula Weight 144.25
MDL Number MFCD00046585
Quantity 50g
Synonym Ethyl methyl 2-methylbutyl carbinol

Applications
It is used in the synthesis of tertiary 14C-labelled nonylphenol isomers..

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.