Please login to your online account to display your discounted pricing

3,5-Dimethyl-1H-pyrrole-2-carbaldehyde, ≥95%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC18004CB

 View more versions of this product

Catalog No. CC18004CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 123.15
Quantity 250mg
Assay 95%
CAS Max % 100.0
Chemical Name or Material 3, 5-Dimethyl-1H-pyrrole-2-carbaldehyde
Assay Percent Range 95%
Molecular Formula C7H9NO
Linear Formula Yes
Packaging Amber glass bottle
MDL Number MFCD00111522
CAS 2199-58-8
CAS Min % 94.5

Specifications

Chemical Name or Material 3, 5-Dimethyl-1H-pyrrole-2-carbaldehyde
CAS 2199-58-8
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H9NO
Linear Formula Yes
Formula Weight 123.15
MDL Number MFCD00111522