missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde, ≥95%, Thermo Scientific™
Chemical Identifiers
CAS | 2199-58-8 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.155 |
MDL Number | MFCD00111522 |
InChI Key | RDFZYUOHJBXMJA-UHFFFAOYSA-N |
Synonym | 3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole |
PubChem CID | 270465 |
IUPAC Name | 3,5-dimethyl-1H-pyrrole-2-carbaldehyde |
SMILES | CC1=CC(=C(N1)C=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC18004CB
|
Thermo Scientific Chemicals
CC18004CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC18004DA
|
Thermo Scientific Chemicals
CC18004DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC18004DE
|
Thermo Scientific Chemicals
CC18004DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
2199-58-8 | |
123.155 | |
RDFZYUOHJBXMJA-UHFFFAOYSA-N | |
270465 | |
CC1=CC(=C(N1)C=O)C |
C7H9NO | |
MFCD00111522 | |
3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole | |
3,5-dimethyl-1H-pyrrole-2-carbaldehyde |
Specifications
2199-58-8 | |
100.0 | |
Amber glass bottle | |
Yes | |
250mg | |
RDFZYUOHJBXMJA-UHFFFAOYSA-N | |
3,5-dimethyl-1H-pyrrole-2-carbaldehyde | |
270465 | |
95% |
94.5 | |
95% | |
C7H9NO | |
MFCD00111522 | |
3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole | |
CC1=CC(=C(N1)C=O)C | |
123.155 | |
123.15 | |
3, 5-Dimethyl-1H-pyrrole-2-carbaldehyde |