missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde, ≥95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC18004DA
Specifications
3, 5-Dimethyl-1H-pyrrole-2-carbaldehyde | |
94.5 | |
95% | |
C7H9NO | |
MFCD00111522 | |
3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole | |
CC1=CC(=C(N1)C=O)C | |
123.155 | |
123.15 |
2199-58-8 | |
100.0 | |
Amber glass bottle | |
Yes | |
1g | |
RDFZYUOHJBXMJA-UHFFFAOYSA-N | |
3,5-dimethyl-1H-pyrrole-2-carbaldehyde | |
270465 | |
95% |
Chemical Identifiers
2199-58-8 | |
123.155 | |
RDFZYUOHJBXMJA-UHFFFAOYSA-N | |
270465 | |
CC1=CC(=C(N1)C=O)C |
C7H9NO | |
MFCD00111522 | |
3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole | |
3,5-dimethyl-1H-pyrrole-2-carbaldehyde |