missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-1H-pyrazole, 99%

Manufacturer:  Alfa Aesar™ A1015718

 View more versions of this product

Catalog No. AAA1015718



Description

Description

Applications
3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI)­ oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.

Solubility
Soluble in water, methanol 0.1 g/mL.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

3,5-Dimethyl-1H-pyrazole
105°C to 108°C
6.7
C5H8N2
106325
SDXAWLJRERMRKF-UHFFFAOYSA-N
3,5-dimethyl-1H-pyrazole
6210
99%
218°C
50g
67-51-6
MFCD00005243
3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256
CC1=CC(=NN1)C
96.133
96.13

Chemical Identifiers

67-51-6
96.133
SDXAWLJRERMRKF-UHFFFAOYSA-N
6210
CC1=CC(=NN1)C
C5H8N2
MFCD00005243
3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256
3,5-dimethyl-1H-pyrazole
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UQ6477600

TSCA : TSCA

SDS Documents