missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5405003

 View more versions of this product

Catalog No. AAH5405003



Description

Description

3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride
C5H7ClN2O2S
1 g
Moisture sensitive
NBGSZCQLPLYKJH-UHFFFAOYSA-N
3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride
594587
97%
80466-78-0
MFCD04969910
UN3261
1h-pyrazole-4-sulfonylchloride, 3,5-dimethyl, 3,5-dimethylpyrazol-4-yl chlorosulfone, 1h-pyrazole-4-sulfonyl chloride, 3,5-dimethyl, pyrazole, 4-chlorosulfonyl-3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-sulfonylchloride, #
CC1=C(C(C)=NN1)S(Cl)(=O)=O
194.63
194.64

Chemical Identifiers

80466-78-0
194.63
NBGSZCQLPLYKJH-UHFFFAOYSA-N
594587
CC1=C(C(C)=NN1)S(Cl)(=O)=O
C5H7ClN2O2S
MFCD04969910
1h-pyrazole-4-sulfonylchloride, 3,5-dimethyl, 3,5-dimethylpyrazol-4-yl chlorosulfone, 1h-pyrazole-4-sulfonyl chloride, 3,5-dimethyl, pyrazole, 4-chlorosulfonyl-3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-sulfonylchloride, #
3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only