Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-1H-pyrazole-1-carboxamide, 98+%

$58.81 - $692.68

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyl-1H-pyrazole-1-carboxamide
CAS 934-48-5
Molecular Formula C6H9N3O
Formula Weight 139.16
MDL Number MFCD00003126
Beilstein 123422
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1452309

  SDS
alfa aesar™
A1452309
10g Each for $58.81

AAA1452318

  SDS
alfa aesar™
A1452318
50g Each for $164.70

AAA1452330

  SDS
alfa aesar™
A1452330
250g Each for $692.68
Description & Specifications

Specifications

Formula Weight 139.16
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole-1-carboxamide
Assay ≥98%
Molecular Formula C6H9N3O
Melting Point 112° to 114°C
MDL Number MFCD00003126
CAS 934-48-5
Beilstein 123422
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.