missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-1H-pyrazole-1-carboxamide, 98+%

Manufacturer:  Alfa Aesar™ A1452318

 View more versions of this product

Catalog No. AAA1452318



Specifications

Specifications

3,5-Dimethyl-1H-pyrazole-1-carboxamide
50g
C6H9N3O
123422
AKRDSDDYNMVKCX-UHFFFAOYSA-N
3,5-dimethylpyrazole-1-carboxamide
70280
≥98%
112°C to 114°C
934-48-5
MFCD00003126
3,5-dimethyl-1h-pyrazole-1-carboxamide, 1h-pyrazole-1-carboxamide, 3,5-dimethyl, 3,5-dimethylpyrazolecarboxamide, acmc-20anc7, timtec-bb sbb004176, 1-carbamoyl-3,5-dimethylpyrazole, 3,5-dimethyl-1-carbamoylpyrazole, 1-carboxamido-3,5-dimethylpyrazole, 3,5-dimethyl-1-pyrazolecarboxamide, 3,5-dimethylpyrazole-1-carboxyamide
CC1=CC(=NN1C(=O)N)C
139.158
139.16

Chemical Identifiers

934-48-5
139.158
AKRDSDDYNMVKCX-UHFFFAOYSA-N
70280
CC1=CC(=NN1C(=O)N)C
C6H9N3O
MFCD00003126
3,5-dimethyl-1h-pyrazole-1-carboxamide, 1h-pyrazole-1-carboxamide, 3,5-dimethyl, 3,5-dimethylpyrazolecarboxamide, acmc-20anc7, timtec-bb sbb004176, 1-carbamoyl-3,5-dimethylpyrazole, 3,5-dimethyl-1-carbamoylpyrazole, 1-carboxamido-3,5-dimethylpyrazole, 3,5-dimethyl-1-pyrazolecarboxamide, 3,5-dimethylpyrazole-1-carboxyamide
3,5-dimethylpyrazole-1-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents