Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  408520250

 View more versions of this product

Catalog No. AC408520250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 200.24
Quantity 25g
TSCA TSCA
EINECS Number 244-750-9
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
MDL Number MFCD00051396
Chemical Name or Material 3, 5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde
Name Note 99%
Assay Percent Range 99%
Molecular Formula C12H12N2O
CAS 22042-79-1

Specifications

Chemical Name or Material 3, 5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde
Name Note 99%
CAS 22042-79-1
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C12H12N2O
Formula Weight 200.24
MDL Number MFCD00051396
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents