Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethyl-1-phenyl-1H-pyrazole, 97%

$57.57 - $233.55

Chemical Identifiers

Chemical Name or Material 3,5-Dimethyl-1-phenyl-1H-pyrazole
CAS 1131-16-4
Molecular Formula C11H12N2
Formula Weight 172.23
MDL Number MFCD00020728
Beilstein 132902
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2548906

  SDS
alfa aesar™
B2548906
5g Each for $57.57

AAB2548914

  SDS
alfa aesar™
B2548914
25g Each for $233.55
Description & Specifications

Specifications

Formula Weight 172.23
Chemical Name or Material 3,5-Dimethyl-1-phenyl-1H-pyrazole
TSCA TSCA
Assay 97%
Molecular Formula C11H12N2
Boiling Point 144° to 145°C (12mmHg)
MDL Number MFCD00020728
CAS 1131-16-4
Beilstein 132902
Quantity 5g
Flash Point >110°C (230°F)