missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, ≥97%, Maybridge

Manufacturer:  Fisher Scientific Company CC48003DA

 View more versions of this product

Catalog No. CC48003DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

270.74
1g
80.0
3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
97+%
C11H11ClN2O2S
Technical Grade
Tech.
100.0
MFCD04968669
59340-26-0

Chemical Identifiers

59340-26-0
100.0
C11H11ClN2O2S
MFCD04968669
Tech.
80.0
97+%
270.74
3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.