Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, ≥97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC48003CB

 View more versions of this product

Catalog No. CC48003CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 270.74
Quantity 250mg
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C11H11ClN2O2S
CAS 59340-26-0
MDL Number MFCD04968669
Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
CAS 59340-26-0
CAS Min % 80.0
CAS Max % 100.0
Assay Tech.
Assay Percent Range 97+%
Molecular Formula C11H11ClN2O2S
Formula Weight 270.74
MDL Number MFCD04968669
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents